missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium L-3-Phenyl-d5-lactate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Sodium L-3-Phenyllactate-D5 (phenyl-D5) |
| Molecular Formula | C9 D5 H4 O3 . Na |
| Quantity | 0.01 g |
| Synonym | Sodium (αS)-α-hydroxybenzene-d5-propanoate, Sodium (S)-3-(phenyl-d5)lactate, L-3-Phenyllactic-d5 acid sodium salt, Sodium L-3-phenyllactate-D5 (phenyl-D5), (αS)-α-Hydroxybenzene-d5-propanoic acid sodium salt, Sodium L-3-phenyl-d5-lactate |
| InChI Formula | InChI=1 S/C9H10O3.Na/c10-8(9(11)12)6-7-4-2-1-3-5-7;/h1-5,8,10 H,6H2,(H,11,12);/q;+1/p-1/t8-;/m0./s1/i1D,2 D,3 D,4 D,5 D; |
| SMILES | [Na+].[2 H]c1c([2 H])c([2 H])c(C[C@H](O)C(=O)[O-])c([2 H])c1[2 H] |
| IUPAC Name | sodium;(2 S)-2-hydroxy-3-(2,3,4,5,6-pentadeuteriophenyl)propanoate |
| Molecular Weight (g/mol) | 193.186 |
| Formula Weight | 193.076 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction