missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metabisulfite, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10736111 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Unit Size:
1kg
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10736111 1 kg 1kg
10438420 500 g 500g
10223970 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10736111 Supplier Fisher Chemical Supplier No. S/5240/60

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium Metabisulfite, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Melting Point 150°C
Color White
pH 4 to 6
Physical Form Powder/Solid
Quantity 1 kg
Formula Weight 190.1g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Sodium metabisulfite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.