Learn More
Sodium Naphthalene-2,6-disulfonate, TRC
Specifications
Specifications
| Chemical Name or Material | Sodium Naphthalene-2,6-disulfonate |
| CAS | 1655-45-4 |
| Molecular Formula | C10H6Na2O6S2 |
| Quantity | 50 g |
| Synonym | 2,6-Naphthalenedisulfonic acid','sodium salt (1:2) (ACI), 2,6-Naphthalenedisulfonic acid','disodium salt (8CI','9CI), Disodium 2,6-naphthalenedisulfonate, Sodium naphthalene-2,6-disulfonate, 2,6-Naphthalenedisulfonic acid disodium salt |
| InChI Formula | InChI=1S/C10H8O6S2.2Na/c11-17(12,13)9-3-1-7-5-10(18(14,15)16)4-2-8(7)6-9;;/h1-6H,(H,11,12,13)(H,14,15,16);;/q;2*+1/p-2 |
| SMILES | [Na+].[Na+].[O-]S(=O)(=O)c1ccc2cc(ccc2c1)S(=O)(=O)[O-] |
| IUPAC Name | disodium;naphthalene-2,6-disulfonate |
| Molecular Weight (g/mol) | 332.26 |
| Formula Weight | 331.9401 |