missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Octanoate-8,8,8-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Sodium Octanoate-8,8,8-D3 |
| CAS | 1219795-01-3 |
| Molecular Formula | C8H12D3NaO2 |
| Quantity | 0.25 g |
| Synonym | Sodium Octanoate-8,8,8-d3, Sodium octanoate-8,8,8-d15, Sodium octanoate-8,8,8-D3, Octanoic-8,8,8-d3 acid sodium salt, Octanoic acid 8,8,8-D3 sodium salt, Octanoic acid-d3 sodium salt, Caprylic acid-ω-d3 sodium salt, Sodium caprylate-ω-d3 |
| InChI Formula | InChI=1 S/C8H16O2.Na/c1-2-3-4-5-6-7-8(9)10;/h2-7H2,1H3,(H,9,10);/q;+1/p-1/i1D3; |
| SMILES | [O-]C(CCCCCCC([2 H])([2 H])[2 H])=O.[Na+] |
| IUPAC Name | sodium octanoate-8,8,8-d3 |
| Molecular Weight (g/mol) | 169.21 |
| Formula Weight | 169.12 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction