missing translation for 'onlineSavingsMsg'
Learn More

Sodium Orthovanadate, TRC

Product Code. 30426146
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Unit Size:
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426146 100 g 100g
30414992 5 g -
30414398 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426146 Supplier Toronto Research Chemicals Supplier No. TRCS655003100G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13721-39-6
Molecular Formula Na3O4V
Molecular Weight (g/mol) 183.91
Synonym NSC 79534, Sodium o-Vanadate, Sodium Orthovanadate, Sodium Orthovanadate (Na3VO4), Sodium Tetraoxovanadate(3-), Sodium Vanadate, Sodium Vanadate (Na3VO4), Trisodium Orthovanadate, Trisodium Vanadate, Sodium Vanadate(V) (Na3VO4)
IUPAC Name trisodium;trioxido(oxo)vanadium
SMILES [Na+].[Na+].[Na+].[O-][V](=O)([O-])[O-]

Specifications

Quantity 100 g
InChI Formula InChI=1S/3Na.4O.V/q3*+1;;3*-1;
Formula Weight 183.8929
Chemical Name or Material Sodium Orthovanadate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.