missing translation for 'onlineSavingsMsg'
Learn More

Sodium Perchlorate, TRC

Product Code. 30137790
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137790 100 g 100g
17958539 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 30137790 Supplier Toronto Research Chemicals Supplier No. TRCS655060100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-89-0
Molecular Formula Cl O4 . Na
Molecular Weight (g/mol) 122.44
Synonym Perchloric acid, sodium salt
IUPAC Name sodium;perchlorate
SMILES [Na+].[O-]Cl(=O)(=O)=O

Specifications

Quantity 100 g
InChI Formula InChI=1S/ClHO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1
Formula Weight 121.938
Chemical Name or Material Sodium perchlorate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.