missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phenyl Phosphate Dibasic Dihydrate, TRC

Product Code. 30426690
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426690 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426690 Supplier Toronto Research Chemicals Supplier No. TRCS67327010G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Phenyl Phosphate Dibasic Dihydrate
CAS 66778-08-3
Molecular Formula C6H5Na2O4P • 2(H 2O)
Quantity 10 g
Synonym Phosphoric Acid','Monophenyl Ester','Sodium Salt','Hydrate (1:2:2)
InChI Formula InChI=1S/C6H7O4P.2Na.2H2O/c7-11(8,9)10-6-4-2-1-3-5-6;;;;/h1-5H,(H2,7,8,9);;;2*1H2/q;2*+1;;/p-2
SMILES O=P(O[Na])(O[Na])OC1=CC=CC=C1.O.O
IUPAC Name sodium phenyl phosphate dihydrate
Molecular Weight (g/mol) 254.08
Formula Weight 253.99
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.