missing translation for 'onlineSavingsMsg'
Learn More

Melford Sodium Phosphate, Monobasic, Anhydrous

Product Code. 30018793
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018793 5 kg 5kg
30018791 1 kg 1kg
30018792 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018793 Supplier Melford Supplier No. S231855000.0
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Sodium Phosphate, Monobasic, Anhydrous is a chemical compound that plays a significant role in the life science research.

It is also known as monosodium phosphate or sodium dihydrogen phosphate. The anhydrous designation indicates that it is the form of the compound without any water molecules.

In the life science field, it is primarily utilized as a buffering agent and as a source of phosphate ions. Buffers are substances that help maintain a stable pH in biological and biochemical processes, ensuring optimal conditions for enzymatic reactions and other cellular activities.

Sodium Phosphate, Monobasic, Anhydrous is often used in laboratory settings for the preparation of various biological reagents, cell culture media, and buffer solutions. It can be combined with other phosphate compounds to create buffer systems with specific pH ranges and buffering capacities, making it suitable for a wide range of experimental conditions

Moreover, the compound is used in molecular biology techniques like DNA and protein electrophoresis, as it provides the necessary pH control for separation and analysis of biomolecules. It is also employed in the preparation of phosphate-based saline solutions.

Specifications

Chemical Name or Material Sodium Phosphate, Monobasic, Anhydrous
CAS 7558-80-7
Quantity 5 kg
Molecular Formula NaH2PO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.