missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Potassium ATPase Alpha 2 Antibody (SR1026), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22392-100ul
This item is not returnable.
View return policy
Description
Sodium Potassium ATPase Alpha 2 Monoclonal antibody specifically detects Sodium Potassium ATPase Alpha 2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Sodium Potassium ATPase Alpha 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1026 | |
| Western Blot 1:500-1:2000 | |
| alpha-A(+) catalytic polypeptide, ATPase, Na+/K+ transporting, alpha 2 (+) polypeptide, ATPase, Na+/K+ transporting, alpha 2 polypeptide, EC 3.6.3, EC 3.6.3.9, KIAA0778, MGC59864, MHP2, migraine, hemiplegic 2, Na(+)/K(+) ATPase alpha-2 subunit, Na+/K+ ATPase, alpha-B polypeptide, Sodium pump subunit alpha-2, sodium/potassium-transporting ATPase alpha-2 chain, sodium/potassium-transporting ATPase subunit alpha-2, sodium-potassium ATPase | |
| A synthesized peptide derived from human Sodium Potassium ATPase Alpha 2 (Uniprot #: P50993) | |
| 100 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 477 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction