missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Potassium ATPase Beta 1 Antibody (S02-8B6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20085-50ul
This item is not returnable.
View return policy
Description
Sodium Potassium ATPase Beta 1 Monoclonal antibody specifically detects Sodium Potassium ATPase Beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Sodium Potassium ATPase Beta 1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:20-1:50, Immunohistochemistry-Paraffin | |
| adenosinetriphosphatase, ATP1B, ATPase, Na+/K+ transporting, beta 1 polypeptide, Beta 1-subunit of Na(+), K(+)-ATPase, MGC1798, Na, K-ATPase beta-1 polypeptide, sodium/potassium-dependent ATPase beta-1 subunit, Sodium/potassium-dependent ATPase subunit beta-1, sodium/potassium-transporting ATPase beta-1 chain, sodium/potassium-transporting ATPase subunit beta-1 | |
| A synthetic peptide of human Sodium Potassium ATPase Beta 1 (Uniprot # P05026) | |
| 50 μg | |
| Cancer, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers, Signal Transduction | |
| 481 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S02-8B6 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction