missing translation for 'onlineSavingsMsg'
Learn More

Sodium Pyrophosphate Decahydrate, TRC

Product Code. 30414717
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414717 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414717 Supplier Toronto Research Chemicals Supplier No. TRCS083925250G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Pyrophosphate Decahydrate
CAS 13472-36-1
Quantity 250 g
Molecular Formula Na4O7P2 . 10H2O
Synonym Tetrasodium Pyrophosphate Decahydrate, Sodium Diphosphate Decahydrate
InChI Formula InChI=1S/4Na.H4O7P2.10H2O/c;;;;1-8(2,3)7-9(4,5)6;;;;;;;;;;/h;;;;(H2,1,2,3)(H2,4,5,6);10*1H2/q4*+1;;;;;;;;;;;/p-4
SMILES O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P(=O)([O-])OP(=O)([O-])[O-]
IUPAC Name tetrasodium;phosphonato phosphate;decahydrate
Molecular Weight (g/mol) 446.06
Formula Weight 445.9767
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.