missing translation for 'onlineSavingsMsg'
Learn More

Sodium (R)-3-(1-Hydroxy-2-(methylamino)ethyl)phenyl Sulfate, TRC

Product Code. 30422934
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30422934 1 mg 1mg
30421844 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30422934 Supplier Toronto Research Chemicals Supplier No. TRCS6349601MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium (R)-3-(1-Hydroxy-2-(methylamino)ethyl)phenyl Sulfate
Molecular Formula C9H12NNaO5S
Quantity 1 mg
Synonym (R)-Phenylephrine 3-O-Sulfate sodium salt
InChI Formula InChI=1S/C9H13NO5S.Na/c1-10-6-9(11)7-3-2-4-8(5-7)15-16(12,13)14;/h2-5,9-11H,6H2,1H3,(H,12,13,14);/q;+1/p-1/t9-;/m0./s1
SMILES [Na+].CNC[C@H](O)c1cccc(OS(=O)(=O)[O-])c1
Molecular Weight (g/mol) 269.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.