missing translation for 'onlineSavingsMsg'
Learn More

Melford Sodium Sulfate, Anhydrous

Product Code. 30018808
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018808 1 kg 1kg
30018809 500 g 500g
30018810 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018808 Supplier Melford Supplier No. S251501000.0
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Fine white granular form.

Fine white granular form. Store at room temperature.

Specifications

Chemical Name or Material Sodium Sulfate, Anhydrous
CAS 7757-82-6
Quantity 1 kg
Molecular Formula Na2SO4
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.