missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tetraborate, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Disodium tetraborate |
| CAS | 1330-43-4 |
| Molecular Formula | B4 O7 . 2Na |
| Quantity | 25 g |
| Synonym | Sodium tetraborate, Boron sodium oxide (B4Na2O7) (9CI','ACI), Boric acid (H2B4O7)','disodium salt (8CI), Sodium tetraborate (Na2B4O7) (7CI), Anhydrous borax, Borax glass, DC Dehybor, Dehybor, Ecoboron PRO, Etibor 68, Fireless B, Fireless B Liquid, FR 28, FR 28 (borate), Fused Borax, Granubor, Rasorite 65, Sodium biborate, Sodium borate, Sodium boron oxide (Na2B4O7), Spraybor |
| InChI Formula | InChI=1S/B4O7.2Na/c5-1-7-3-9-2(6)10-4(8-1)11-3;;/q-2;2*+1 |
| SMILES | [Na+].[Na+].[O-]B1OB2OB([O-])OB(O1)O2 |
| IUPAC Name | disodium;3,7-dioxido-2,4,6,8,9-pentaoxa-1,3,5,7-tetraborabicyclo[3.3.1]nonane |
| Molecular Weight (g/mol) | 201.22 |
| Formula Weight | 201.9812 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction