missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, 98 to 102% (titration), Reagent Grade, Honeywell™

Product Code. 15658110 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
3 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
3kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15658110 3 kg Poly bottle 3kg
15648110 250 g Poly bottle 250g
15638110 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15658110 Supplier Honeywell Research Chemicals Supplier No. S77573KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 98-102% (titration)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
SMILES [Na+].[S-]C#N

Specifications

Melting Point 308°C
Color Colorless
Physical Form Solid
Quantity 3 kg
Linear Formula NaSCN
UN Number NONH for all modes of transport
Beilstein 3594965
IUPAC Name sodium;thiocyanate
Formula Weight 81.07g/mol
Percent Purity 98 to 102% (titration)
Odor Odorless
Packaging Poly bottle
Density 1,76 g/cu.cm at 20°C
Chemical Name or Material Sodium thiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.