missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15508194 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15508194 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15508194 Supplier Avantor J.T.Baker Supplier No. 393801

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 540-72-7
Physical Form Crystals
Quantity 500 g
Molecular Formula CNNaS
MDL Number MFCD00011123
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
SMILES [Na+].[S-]C#N
IUPAC Name sodium;thiocyanate
Molecular Weight (g/mol) 81.07
PubChem CID 516871
ChEBI CHEBI:30952
Formula Weight 81.07
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.00kg
Chemical Name or Material Sodium Thiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.