missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.005M, Chem-Lab

Product Code. 11979587
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11979587 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11979587 Supplier Chem-Lab Supplier No. CL05.1441.1000
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used as a redox titrant in iodometry for TMAR chlorine determination according to SERES. Sodium Thiosulfate, 0.005M, Chem-Lab is a solution of 1.241g Na2S2O3·5H2O per 1L H2O. It is an inorganic compound with an excellent shelf life.

The thiosulfate anion reacts stoichiometrically with iodine in aqueous solution. Due to the quantitative nature of this reaction, sodium thiosulfate is used as a redox titrant in volumetric analysis and is a component of iodine clock experiments. It can be used to measure the oxygen content of water and to estimate the concentration of certain compounds in solution (hydrogen peroxide, chlorine). It also has many medicinal uses.

Specifications

Boiling Point 100°C
Melting Point 48.3°C
CAS 10102-17-7, 7732-18-5
Physical Form Liquid
Quantity 1 L
Molecular Formula H10Na2O8S2
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
Solubility Information Soluble in water
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
Molecular Weight (g/mol) 248.172
PubChem CID 61475
ChEBI CHEBI:32150
Formula Weight 248.18g/mol
Concentration or Composition (by Analyte or Components) 0.005mol/L
Density 1.00g/mL
Chemical Name or Material Sodium Thiosulfate Pentahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.