missing translation for 'onlineSavingsMsg'
Learn More

Sodium Triphosphate, TRC

Product Code. 30427306
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427306 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427306 Supplier Toronto Research Chemicals Supplier No. TRCS2243505G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Triphosphate
CAS 7758-29-4
Quantity 5 g
Molecular Formula 5 Na . O10 P3
Synonym Pentasodium Triphosphate, Pentasodium Triphosphate (Na5P3O10), Pentasodium Tripolyphosphate, Rhodiaphos A 4010, Rhodiaphos A 70HG, Rhodiaphos C 90H, Sodium Metaphosphate Oxide (Na5(PO3)3O), Sodium Phosphate (Na5P3O10), Sodium Polyphosphate (Na5P3O10), Sodium Triphosphate, Thermphos L 50, Thermphos L 500, Thermphos N, Thermphos NW
InChI Formula InChI=1S/5Na.H5O10P3/c;;;;;1-11(2,3)9-13(7,8)10-12(4,5)6/h;;;;;(H,7,8)(H2,1,2,3)(H2,4,5,6)/q5*+1;/p-5
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[O-]P(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-]
IUPAC Name pentasodium;[oxido(phosphonatooxy)phosphoryl] phosphate
Molecular Weight (g/mol) 367.864
Formula Weight 367.819
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.