missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tripolyphosphate, 85%, Technical grade, Honeywell Fluka™

Product Code. 15692380 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15692380 500 g Poly Bottle 500g
15682380 25 g Poly Bottle 25g
15672380 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15692380 Supplier Honeywell-Fluka Supplier No. 238503500G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Technical Grade, 85%

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-29-4
Molecular Formula Na5O10P3
Molecular Weight (g/mol) 367.86
MDL Number MFCD00003514
InChI Key HWGNBUXHKFFFIH-UHFFFAOYSA-I
Synonym sodium tripolyphosphate, sodium triphosphate, pentasodium triphosphate, stpp, thermphos, armofos, polygon, tripoly, rhodiaphos lv, thermphos n
PubChem CID 24455
IUPAC Name pentasodium;[oxido(phosphonatooxy)phosphoryl] phosphate
SMILES [O-]P(=O)([O-])OP(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+].[Na+]

Specifications

Melting Point 622°C
Color White
Quantity 500 g
Linear Formula Na5P3O10
UN Number NONH for all modes of transport
Formula Weight 367.86g/mol
Percent Purity 85%
Grade Technical
Packaging Poly Bottle
Appearance Granular powder
Density 2.52g/cm3
Chemical Name or Material Sodium Tripolyphosphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.