missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SOGA1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA572776

Product Code. 15941435

  • 520.00€ / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SOGA1 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

SOGA1 regulates autophagy by playing a role in the reduction of glucose production in an adiponectin- and insulin-dependent manner.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SOGA1
Polyclonal
Unconjugated
Soga1
80-kDa SOGA fragment; 9830001H06Rik; AI426038; C20orf117; C-terminal 80 kDa form; D430036N24Rik; dJ132F21.1; KIAA0889; N-terminal form; Protein SOGA1; Soga; SOGA family member 1; Soga1; Suppressor of glucose by autophagy; suppressor of glucose from autophagy; suppressor of glucose, autophagy associated 1; suppressor of glucose, autophagy-associated protein 1
Rabbit
Antigen affinity chromatography
RUO
140710
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.02% sodium azide
O94964
Soga1
SOGA1 antibody for a 16 amino acid peptide near the carboxy terminus of human SOGA. The immunogen is within the last 50 amino acids of SOGA1.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.