missing translation for 'onlineSavingsMsg'
Learn More

Solabegron Hydrochloride, TRC

Product Code. 30420935
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30420935 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420935 Supplier Toronto Research Chemicals Supplier No. TRCS67651010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Solabegron Hydrochloride
CAS 451470-34-1
Molecular Formula C23H23ClN2O3.HCl
Quantity 10 mg
Synonym 3'-[[2-[[(2R)-2-(3-Chlorophenyl)-2-hydroxyethyl]amino]ethyl]amino]-[1,1'-biphenyl]-3-carboxylic Acid Hydrochloride, 3'-[[2-[[(2R)-2-(3-Chlorophenyl)-2-hydroxyethyl]amino]ethyl]amino]-[1,1'-biphenyl]-3-carboxylic Acid Monohydrochloride, (R)-3'-((2-((2-(3-Chlorophenyl)-2-hydroxyethyl)amino)ethyl)amino)-[1,1'-biphenyl]-3-carboxylic Acid Hydrochloride, GW 427353B
InChI Formula InChI=1S/C23H23ClN2O3.ClH/c24-20-8-2-6-18(13-20)22(27)15-25-10-11-26-21-9-3-5-17(14-21)16-4-1-7-19(12-16)23(28)29;/h1-9,12-14,22,25-27H,10-11,15H2,(H,28,29);1H/t22-;/m0./s1
SMILES Cl.O[C@@H](CNCCNc1cccc(c1)c2cccc(c2)C(=O)O)c3cccc(Cl)c3
IUPAC Name 3-[3-[2-[[(2R)-2-(3-chlorophenyl)-2-hydroxyethyl]amino]ethylamino]phenyl]benzoic acid;hydrochloride
Molecular Weight (g/mol) 447.4
Formula Weight 446.1164
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.