missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A H5N8 HA (A/Astrakhan/3212/2020) Recombinant Rabbit Monoclonal Antibody (HL1550)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA5-47093
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Specifications
| Influenza A H5N8 HA (A/Astrakhan/3212/2020) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL1550 | |
| Unconjugated | |
| flu hemagglutinin; influenza hemagglutinin | |
| Recombinant protein encompassing a sequence within the center region of Influenza A virus H5N8 HA (Hemagglutinin)(A/Astrakhan/3212/2020)). The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction