Learn More
- Home
- Products
- Stoppers and Closures
- Stoppers
- Fisherbrand™ Solid Neoprene Rubber Bungs
Fisherbrand™ Solid Neoprene Rubber Bungs
Product Code. 11597093 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Diameter (Metric) Bottom:
3 mm
4 mm
6 mm
7 mm
9 mm
10 mm
11 mm
13 mm
17 mm
19 mm
25 mm
27 mm
33 mm
39 mm
43 mm
49 mm
53 mm
57 mm
62 mm
67 mm
71 mm
75 mm
79 mm
86 mm
92 mm
99 mm
Diameter (Metric) Top:
4.5 mm
5.8 mm
8 mm
9 mm
11.5 mm
12.5 mm
14 mm
16 mm
20.3 mm
22.5 mm
28.5 mm
31 mm
38.5 mm
44.5 mm
49 mm
55 mm
59.5 mm
64 mm
69 mm
74 mm
78 mm
85 mm
89 mm
96 mm
102 mm
109 mm
Length (Metric):
12 mm
14 mm
16 mm
20 mm
24 mm
26 mm
28 mm
32 mm
38.5 mm
42 mm
45.5 mm
49 mm
54 mm
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Pack of 50
28 product options available for selection
| Product Code. | Diameter (Metric) Bottom | Diameter (Metric) Top | Length (Metric) | Unit Size |
|---|---|---|---|---|
| 11597093 | 19 mm | 22.5 mm | 28 mm | Pack of 10 |
| 11547093 | 10 mm | 12.5 mm | 20 mm | Pack of 20 |
| 11587093 | 17 mm | 20.3 mm | 26 mm | Pack of 20 |
| 11557093 | 11 mm | 14 mm | 24 mm | Pack of 20 |
| 11527103 | 25 mm | 28.5 mm | 28 mm | Pack of 10 |
| 11537103 | 27 mm | 31 mm | 32 mm | Pack of 10 |
| 11567103 | 33 mm | 38.5 mm | 38.5 mm | Pack of 5 |
| 11597103 | 39 mm | 44.5 mm | 38.5 mm | Pack of 5 |
| 11517113 | 43 mm | 49 mm | 42 mm | Pack of 2 |
| 11537113 | 49 mm | 55 mm | 42 mm | Pack of 2 |
| 11587083 | 4 mm | 5.8 mm | 14 mm | Pack of 50 |
| 11547113 | 53 mm | 59.5 mm | 45.5 mm | Each |
| 11557113 | 57 mm | 64 mm | 49 mm | Each |
| 11567113 | 62 mm | 69 mm | 49 mm | Each |
| 11577113 | 67 mm | 74 mm | 49 mm | Each |
| 11507093 | 6 mm | 8 mm | 16 mm | Pack of 20 |
| 11587113 | 71 mm | 78 mm | 49 mm | Each |
| 11597113 | 75 mm | 85 mm | 54 mm | Each |
| 11517093 | 7 mm | 9 mm | 16 mm | Pack of 20 |
| 11537093 | 9 mm | 11.5 mm | 20 mm | Pack of 20 |
| 11507123 | 79 mm | 89 mm | 54 mm | Each |
| 11517123 | 86 mm | 96 mm | 54 mm | Each |
| 11527123 | 92 mm | 102 mm | 54 mm | Each |
| 11537123 | 99 mm | 109 mm | 54 mm | Each |
| 11577083 | 3 mm | 4.5 mm | 12 mm | Pack of 50 |
| 10587064 | 3 mm | 4.5 mm | 12 mm | Pack of 50 |
| 11567093 | 13 mm | 16 mm | 24 mm | Pack of 20 |
| 10265114 | 13 mm | 16 mm | 24 mm | Pack of 20 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
Specifica
Specifica
| For Use With (Application) | For laboratory use |
| Hardness | 46/55 IRHD |
| Length (Metric) | 28 mm |
| Quantity | 10/Pk. |
| Color | Gray |
| Diameter (Metric) Bottom | 19 mm |
| Diameter (Metric) Top | 22.5 mm |
| Material | Neoprene |
| Product Type | Bung |
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?