Learn More
Sonoco™ Utek™ Mailing Refrigerant Packs
Product Code. 11904658
Change view
Click to view available options
Weight (English):
8 oz.
Unit Size:
Pack of 36
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (English) Unit Size
11904658 8 oz. Pack of 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11904658 Supplier Sonoco™ Supplier No. 432
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Phase change materials are ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or CO2
  • Gel packs have a laminated, metalized film exterior that provides radiant heat barrier, resists punctures
  • Single packets are ideal for use as a cubed ice substitute
  • Mat is well suited for wraparound applications
TRUSTED_SUSTAINABILITY
Specifications
Quantity 36 Cs.
Product Type U-tek Refrigerant Pack
Height (English) 0.75 in.
For Use With (Application) Ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or Carbon dioxide
Freezing Point -23°C
Height (Metric) 1.9 cm
Length (English) 6 in.
Length (Metric) 15.2 cm
Material Laminated, Metallized Film
Weight (English) 8 oz.
Weight (Metric) 227 g
Width (English) 4 in.
Width (Metric) 10 cm
Dimensions (L x W x H) 6 x 4 x 0.75 in.