Learn More
Cytiva SOURCE™ 30Q Chromatography Media
Product Code. 11510814 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11510814 50mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11510814 Supplier Cytiva Supplier No. 17127501
Polymeric, strong anion exchanger designed for intermediate purification and large-scale polishing of proteins, peptides and oligonucleotides
  • Mono-sized 30μm rigid polystyrene/divinyl benzene polymer matrix, modified with quaternary ammonium (Q) strong anion exchange groups
  • Low back pressure and high flow rates for high productivity
  • High chemical stability for wide range of working conditions
TRUSTED_SUSTAINABILITY
Specifications
Buffer All common ion exchange Buffers
Product Type Source 30Q chromato graphy media
Quantity 50mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate 2000cm/hr.
Ligand Type Quaternary Ammonium
Max. Pressure 145psi
pH Range 2 to 12
Particle Size 30μm
Support Type Polystyrene/Divinylbenzene
Content And Storage 4° to 30°C
For Use With (Application) Purification and large-scale polishing
Matrix 15Q Chromatography Media
Show More Show Less
compliance-icons
Product Identifier
  • SOURCE 30Q, 50 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol