Learn More
SOX10 Antibody (CL4455) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59050R
Product Code. 30293698
  • 780.00€ / 0.10mL
Description
SOX10 Monoclonal antibody specifically detects SOX10 in Feline,Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SOX10
Monoclonal
DyLight 550
MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of
This antibody was developed using a recombinant protein derived from P56693, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Feline, Human, Mouse
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence
CL4455
50mM Sodium Borate
Mouse
Protein A purified
RUO
6663
Store at 4⪚C in the dark.