Learn More
SOX11 Antibody (CL0143) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52865AF532
Product Code. 30291522
  • 737.00€ / 0.10mL
Description
SOX11 Monoclonal antibody specifically detects SOX11 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SOX11
Monoclonal
Alexa Fluor 532
SRY (sex determining region Y)-box 11, SRY (sex-determining region Y)-box 11, SRY-related HMG-box gene 11, transcription factor SOX-11
This antibody was developed using a recombinant protein derived from P35716, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0143
50mM Sodium Borate
Mouse
Protein A purified
RUO
6664
Store at 4⪚C in the dark.
IgG2a