missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SOX17 Monoclonal antibody specifically detects SOX17 in Human samples. It is validated for Immunocytochemistry
Specifications
Specifications
| Antigen | SOX17 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 614013 |
| Conjugate | DyLight 594 |
| Dilution | Immunocytochemistry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3 |
| Host Species | Mouse |
| Immunogen | E. coli-derived recombinant human SOX17, Asp177-Val414, Accession # Q9H6I2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?