Learn More
Witeg™ Soxhlet Extractor Bulb
Product Code. 11817700
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11817700 125 mL Each
11807700 60 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11817700 Supplier Witeg™ Supplier No. 2 212 125
  • ONLINE EXCLUSIVE PRICE
    114.00€
    99.43€ / Pack of 1
    Save 14.57€
    12% Off
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
For Use With (Equipment) Soxhlet Extraction Equipment
Product Type Soxhlet Extractor Bulb
Capacity (Metric) 125 mL