missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Soybean Oil, TRC
Click to view available options
Quantity:
25 mL
Unit Size:
25mL
Specifications
Specifications
| Chemical Name or Material | Soybean Oil |
| CAS | 8001-22-7 |
| Quantity | 25 mL |
| Synonym | Soy Bean Oil, Soy-bean oil','AMCO','Best One Soya, Bionatrol','CAP 18, CT 7000, Degummed soybean oil, Destiny HC, HY 3050, Imperial Oil, K 1000, Kofa oil, Lipovenos, Nisshin Soybean White Oil, Pro Tech Natur'l Spray Oil, RBD, S-RBD, Shogun C, Soya fat, Vegetoil, Wesson |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction