missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SP 100030

Description
SP 100030 is a potent dual inhibitor of NF-κB and AP-1 transcriptional activity (IC50 = 50 nM). Blocks production of IL-2, IL-8 and TNF-α from Jurkat T cells. Inhibits cytokine production selectively in T cells; exhibits minimal inhibition of cytokine production in other cell types. Decreases arthritic severity in a mouse model of collagen-induced arthritis. Also ameliorates muscle wasting in a rat model of cancer cachexia.
Specifications
Specifications
| Chemical Name or Material | N-[3,5-Bis(trifluoromethyl)phenyl]-2-chloro-4-(trifluoromethyl)-5-pyrimidinecarboxamide |
| CAS | 154563-54-9 |
| Quantity | 10 mg |
| Target | NFkB/IkB Inhibitors |
| Molecular Formula | C14H5ClF9N3O |
| Formula Weight | 437.65 |
| Purity | >99% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?