Learn More
Heidolph™ SP VARIO™ Single-channel Pump Head
Product Code. 10170931 Shop All Heidolph Instruments Products
Change view
Click to view available options
Flow Rate:
2.0 to 4151 mL/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
10170931 2.0 to 4151 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10170931 Supplier Heidolph™ Supplier No. 5234511000
  • ONLINE EXCLUSIVE PRICE
    1130.00€
    968.44€ / Pack of 1
    Save 161.56€
    14% Off
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable for most Heidolph tubing due to adjustable roller spacing
  • Convex rollers do not squeeze or crush organic cell cultures
  • Pump head features ball bearings
  • Rollers made of stainless steel and aluminum
  • Depending on drive and tubing, the flow rate ranges from 2 to 4151 mL/min.
TRUSTED_SUSTAINABILITY
Specifications
Display LED
Flow Rate 2.0 to 4151 mL/min.
IP Rating IP30