missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPA17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13367-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein present at the cell surface. The N-terminus has sequence similarity to human cAMP-dependent protein kinase A (PKA) type II alpha regulatory subunit (RIIa) while the C-terminus has an IQ calmodulin-binding motif. The central portion of the protein has carbohydrate binding motifs and likely functions in cell-cell adhesion. The protein was initially characterized by its involvement in the binding of sperm to the zona pellucida of the oocyte. Recent studies indicate that it is also involved in additional cell-cell adhesion functions such as immune cell migration and metastasis. A retrotransposed pseudogene is present on chromosome 10q22.Specifications
| SPA17 | |
| Polyclonal | |
| Unconjugated | |
| SPA17 | |
| Cancer/testis antigen 22, CT22, SP17, SP17 1, SPA17, sperm autoantigenic protein 17, Sperm protein 17, Sperm surface protein Sp17 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20686, 53340, 85244 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15506, Q62252 | |
| SPA17 | |
| SPA17 Fusion Protein Ag4186 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction