missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15274-1-AP-150UL
This item is not returnable.
View return policy
Description
SPARC is a secreted protein, acidic and rich in cysteines. It is a matrix-associated protein that elicits changes in cell shape, inhibits cell-cycle progression, and influences the synthesis of extracellular matrix (ECM)Specifications
| SPARC | |
| Polyclonal | |
| Unconjugated | |
| SPARC | |
| Basement membrane protein 40, BM 40, ON, Osteonectin, SPARC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20692, 24791, 595124, 6678 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07214, P09486, P16975, P20112 | |
| SPARC | |
| SPARC Fusion Protein Ag7390 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction