Learn More
Heidolph™ Spare Attachment
Product Code. 10724873 Shop All Heidolph Instruments Products
Change view
Click to view available options
Holds:
26 Vessels w/ 10 to 16 mm dia.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
10724873 26 Vessels w/ 10 to 16 mm dia. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10724873 Supplier Heidolph™ Supplier No. 5490160000
  • ONLINE EXCLUSIVE PRICE
    168.00€
    91.00€ / Pack of 1
    Save 77.00€
    45% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with the Multi Reax
Attachment for 26 vessels with diameters from 10 to 16 mm
TRUSTED_SUSTAINABILITY
Specifications
Holds 26 Vessels w/ 10 to 16 mm dia.
Product Type Spare Attachment
For Use With (Equipment) Multi Reax