Learn More
Testo™ Spare Measuring Head with Thermocouple Strip for Temperature Probe
Description
- Replaceable measuring head with thermocouple strip for temperature probe with clamping bracket 0602 4592
- Measuring range: -76 to 266 °F (-60 to +130 °C), briefly up to 536 °F (+280 °C)
- Simply pull out the old measuring head with thermocouple strip and replace with the new measuring head
Specifications
Specifications
| Interface | Plug thermocouple |
| Accuracy | Class 2 (According to standard EN 60584-2, the accuracy of Class 2 refers to -40 to +1200 °C) |
| Temperature Range (Metric) | -60°C to +130°C |
| Length (Metric) | 25.4 mm |