Learn More
SPATA22 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808417
Description
SPATA22 is highly expressed in adult testis. The specific function of SPATA22 is not yet known.Specifications
| SPATA22 | |
| Monoclonal | |
| Unconjugated | |
| SPATA22 | |
| NYD-SP20, NYDSP20 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84690 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2B1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NHS9 | |
| SPATA22 | |
| Full length human recombit protein of human SPATA22 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.