Learn More
SPATA22 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808417S
Description
SPATA22 Monoclonal antibody specifically detects SPATA22 in Human samples. It is validated for Western BlotSpecifications
| SPATA22 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TCG1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57650 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI2B1 | |
| Unconjugated | |
| Cip2a | |
| Gm882; NYDSP20; NYD-SP20; Repro42; RGD1565378; Sgn; Spata22; spermatogenesis associated 22; spermatogenesis-associated protein 22; testicular tissue protein Li 186; testis development protein NYD-SP20; Unknown (protein for MGC:134099) | |
| Human recombinant protein fragment corresponding to amino acids 635-905 of human KIAA1524 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.