Learn More
SPATA2L Monoclonal Antibody (OTI2B4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802370
Description
SPATA2L belongs to the SPATA2 family. The exact function of SPATA2L is not known. There are two different isoforms.Specifications
| SPATA2L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8IUW3 | |
| SPATA2L | |
| Human recombit protein fragment corresponding to amino acids 179-424 of human SPATA2L produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2B4 | |
| Unconjugated | |
| SPATA2L | |
| C16orf76, tamo | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 124044 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.