missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ Absorbents Universal Plus Perforated Chemical Pads

Product Code. 30533369
Change view
Click to view available options
Capacity (English):
23.511 gal.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (English) unitSize
30533369 23.511 gal. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30533369 Supplier Brady™ Supplier No. 813842
Stock Up Your Lab
Use promo code 28007
View all promotions
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

A 2-ply, green, medium weight, bonded and perforated chemical sorbent pad that absorbs chemicals and hazardous liquids. One sided spunbound gives extra strength as well as less lint. Does not degrade or cause a dangerous chemical reaction with the absorbed liquid.

A 2-ply, green, medium weight, bonded and perforated chemical sorbent pad that absorbs chemicals and hazardous liquids.

  • One sided spunbound gives extra strength as well as less lint.
  • Does not degrade or cause a dangerous chemical reaction with the absorbed liquid.
  • Green color alerts workers that the product is being used for hazardous liquids.
  • Green color also allows the user to identify and separate hazardous waste for disposal, resulting in lower disposal costs.
  • Perforated for more flexibility - use only what you need and minimise waste.
  • Perfect to clean up chemical spills or for use in laboratories.

Specifications

Color Green
Capacity (English) 23.511 gal.
Capacity (Metric) 89 L
Type Absorbent Pad
For Use With (Application) Beverage Processing, Chemical, Food Processing, Laboratory, Manufacturing, Maritime, Oil and Gas, Pharmaceutical, Process, Warehouse and Logistics
Model Chemical Sorbent
Length (English) 19.69 in.
Length (Metric) 50 cm
Width (English) 15.74 in.
Width (Metric) 40 cm
Alternate Part Number UN100-E
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.