Learn More
Gestigkeit Harry™ Special Sand For Bath
Product Code. 10290002
Change view
Click to view available options
Quantity:
2 kg
Unit Size:
4kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10290002 2 kg 4kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10290002 Supplier Gestigkeit Harry™ Supplier No. QS4
Edge
For use with universal baths for sand or oil.
  • Provided in quantity of 2kg
  • Allows for even heating
Specifications
Description Special sand for sand bath
Quantity 2 kg