Learn More
Spectrum™ Labs Spectra/Por™ Membrane Kit
Specifications
Specifications
| Product Type | Spectra/Por Membrane Kit |
| Kit Contents | Biotech grade regenerate Cellulose (rc) dialysis tubing in 3 different mwco, 7 spectra/por closures (3 standard, 3 weighted, magnetic-weighted), one spectra/por dialysis reservoir (1800ml) with stir bar |
| Quantity | 1 Kit |
| For Use With (Application) | Perfect solution for dialysis applications |