Learn More
Molecular Devices SpectraMax™ i3x Multi‐Mode Microplate Detection Platform with SoftMax Pro Software
Product Code. 31936260 Shop All Molecular Devices Products
Change view
Click to view available options
Detector Type:
PMT
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Detector Type Unit Size
31936260 PMT 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 31936260 Supplier Molecular Devices Supplier No. I3X
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Perform flexible multi‐mode detection for microplate‐based assays. Supports a wide range of assay workflows with expandable detection capabilities.
  • Designed for multi-mode detection workflows in microplate-based assays
  • Supports a range of detection capabilities for diverse life science applications
  • Enables flexible assay setup with configurable detection parameters
  • Supports biochemical and cell-based assay workflows
  • Provides expandable functionality through modular detection options
  • Integrated with SoftMax Pro Software for data acquisition and analysis
  • Supports scalable workflows for research and laboratory environments
TRUSTED_SUSTAINABILITY
Specifications
Incubator Temperature 4-Zone Incubation to 45°C
Measurement Speed 11 sec. (96 wells), 22 sec. (384 wells)
Shaking Linear, Orbital
Plate Type 6 to 384-well Plates
Detector Type PMT
Software Type SoftMax Pro
Light Source Xenon Flash
Reading Options Endpoint, Kinetic, Spectral Scanning