Learn More
Molecular Devices SpectraMax™ iD5e Multi‐Mode uPlate AS with SoftMax Pro Software
Tienda Molecular Devices ProductosDescripción
- Designed for multi-mode detection workflows in microplate-based assays
- Supports a broad range of detection applications including complex assay workflows
- Enables flexible assay setup with tunable monochromator- and filter-based detection
- Supports biochemical and cell-based assay workflows across life science applications
- Provides enhanced detection capability for advanced and evolving assay requirements
- Includes preconfigured protocols to support common assay workflows
- Integrated with SoftMax Pro Software for data acquisition and analysis
- Designed to support flexible and scalable laboratory workflows
Especificaciones
Especificaciones
| Incubator Temperature | 4-Zone Incubation to 70°C |
| Measurement Speed | 11 sec. (96 wells), 22 sec. (384 wells) |
| Shaking | Linear, Orbital, Double Orbital |
| Plate Type | 6 to 384-well Plates |
| Detector Type | PMT |
| Software Type | SoftMax Pro |
| Light Source | Xenon Flash |
| Reading Options | Endpoint, Kinetic, Spectral Scanning |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?