Learn More
Molecular Devices SpectraMax™ Mini AF System with SoftMax Pro Software
Product Code. 31936333
Change view
Click to view available options
Software Type:
SoftMax Pro
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Software Type Unit Size
31936333 SoftMax Pro 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 31936333 Supplier Molecular Devices Supplier No. SMAX MINI AF
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product is under eco-contribution tax.
Perform fluorescence‐based detection for microplate‐based assays in a compact format. Supports accessible, multi‐mode screening workflows for routine applications.
  • Designed for fluorescence-based detection workflows in microplate assays
  • Supports routine screening applications across life science workflows
  • Provides a compact microplate reader solution for flexible laboratory environments
  • Enables easy assay setup with configurable detection parameters
  • Supports microplate-based workflows for biochemical and cell-based assays
  • Includes preconfigured protocols to support common assay applications
  • Integrated with SoftMax Pro Software for data acquisition and analysis
  • Designed for ease of use and accessible operation in research laboratories
TRUSTED_SUSTAINABILITY
Specifications
Incubator Temperature 4-Zone Incubation to 45°C
Measurement Speed 11 sec. (96 wells), 22 sec. (384 wells)
Shaking Linear, Orbital
Plate Type 6 to 384-well Plates
Detector Type PMT
Software Type SoftMax Pro
Light Source Xenon Flash
Reading Options Endpoint, Kinetic