Learn More
Molecular Devices SpectraMax™ Mini AL System with SoftMax Pro Software
Code produit. 31936339 Tous les produits Molecular Devices Produits
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Software Type:
SoftMax Pro
missing translation for 'unitSize':
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Software Type missing translation for 'unitSize'
31936339 SoftMax Pro 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.
Code produit. 31936339 missing translation for 'mfr' Molecular Devices missing translation for 'supplierNo' SMAX MINI AL
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.
Perform flexible detection for microplate‐based assays in a compact format. Supports routine screening workflows with absorbance and luminescence capabilities.
  • Designed for multi-mode detection workflows in microplate-based assays
  • Supports absorbance and luminescence measurement applications
  • Provides a compact microplate reader solution for flexible laboratory environments
  • Enables easy assay setup with configurable detection parameters
  • Supports microplate-based workflows for biochemical and cell-based assays
  • Includes preconfigured protocols to support common assay applications
  • Integrated with SoftMax Pro Software for data acquisition and analysis
  • Designed for ease of use and accessible operation in research laboratories
TRUSTED_SUSTAINABILITY
Spécification
Shaking Linear, Orbital
Software Type SoftMax Pro