missing translation for 'onlineSavingsMsg'
Learn More

Spectroflux 100, Lithium tetraborate

Product Code. 11346438
Click to view available options
Quantity:
1 kg
5 x 1 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11346438

Brand: Thermo Scientific Alfa Aesar 012078.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 12007-60-2

A general purpose acidic flux for fusing basic oxides, carbonates, alumino-silicates, glass, ceramics, cement steel, aluminum, concrete mixes, soils, magnesites, bauxites and rare earth oxides. Dissolves most refractories. Not suitable for highly acidic samples. Samples must be fully oxidized.Spectroflux 100, Lithium tetraborate is used in enamels. Further, it is a spectroscopic fluxing agent which is utilized for fusing basic oxides, glass, alumino-silicates, ceramics, carbonates, cement, steel, aluminum, soil, concrete mixes, bauxites, magnesites and rare earth oxides. In addition to this, it is used for dissolution of samples containing a high concentration of basic oxides and carbonate rocks and aluminosilicates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A general purpose acidic flux for fusing basic oxides, carbonates, alumino-silicates, glass, ceramics, cement steel, aluminum, concrete mixes, soils, magnesites, bauxites and rare earth oxides. Dissolves most refractories. Not suitable for highly acidic samples. Samples must be fully oxidized.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-60-2

Specifications

Melting Point 930°C
Physical Form Solid
Quantity 1 kg
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Spectroflux 100, Lithium tetraborate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Boron lithium oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.