missing translation for 'onlineSavingsMsg'
Learn More

Spectroflux∣r 1000, Lithium tetraborate, fused, <800 micron, Thermo Scientific Chemicals

Product Code. 11470960
Click to view available options
Quantity:
1 kg
5 x 1 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11470960

Brand: Thermo Scientific Alfa Aesar 096569.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Often used in testing of cement

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Often used in testing of cement
TRUSTED_SUSTAINABILITY

Specifications

CAS 12007-60-2
Physical Form Solid
Quantity 1 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.