Learn More
WTW™ Spectrophotometer photoLab™ 7600 UV-VIS
Product Code. 15345409
Change view
Click to view available options
Product Type:
Spectrophotometer 7600 UV-VIS
Unit Size:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15345409 Spectrophotometer 7600 UV-VIS 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15345409 Supplier WTW™ Supplier No. 250204
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The spectrophotometer photoLab™ 7600 UV-VIS combine routine analysis with spectral analysis and pioneering procedure OptRF. For the standard parameters COD, nitrate and nitrite, a spectrum in the UV range is evaluated by means of complex algorithms which are based on reference spectra. The result is put out directly as concentration reading, without the use of reagents. No potassium dichromate, no digestion - for quick testing.
  • Optical reagent-free measurement of COD, nitrate and nitrite (OptRF)
  • Faster than the fastest digestion
  • No costs, as no reagents or chemicals required
  • Environmentally friendly and no health hazards
  • OptRF-methods are currently being developed for sewage plant outlets
  • Lightweight and powered by car battery, photoLab 7600 UV-VIS can easily be operated on-site 
  • UVT and SAC parameters; for UV-Disinfection as well as for monitoring of organic pollution
  • Five programs with and without turbidity correction are offered 
  • Well-proven routine analysis and wide-ranging application support
  • Simple measuring: photoLab™ 7600 UV-VIS from 190-1100nm supports the fastest and affordable routine analytics by using barcoded test kits and round and rectangular cuvettes just like the VIS version photoLab 7100
  • More than 250 methods are available for water analytics, galvanics and general lab analytics
  • Environmental parameters such as chlorophyll and application procedures for the food and beverage industry are supported by programmed methods and comprehensive programming options 
  • With multi-wavelength, spectral and kinetics measurements, this meter is suitable for mixed applications as well as research
  • PC software photoLab color + Data spectral offer comprehensive color measurement and convenient data management
  • IQ LabLink offers an outstanding connection between IQ Sensor Net and lab measurement for top measurement reliability
  • Measurement data of IQ Sensor Net sensors (VARiON™) can be transferred via USB stick directly into photoLab 7600 UV-VIS and back; faultless and without cable tangle thanks to sensor identification
TRUSTED_SUSTAINABILITY
Specifications
Product Type Spectrophotometer 7600 UV-VIS