Learn More
Spectrum™ Standard RC Ready-to-use Dialysis Sacks Spectra/Por™
Product Code. 15340752
Change view
Click to view available options
MWCO:
12 to 14 kDa
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Unit Size
15340752 12 to 14 kDa Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15340752 Supplier Spectrum™ Supplier No. 132666
  • ONLINE EXCLUSIVE PRICE
    399.00€
    356.19€ / Pack of 10
    Save 42.81€
    10% Off
Estimated Shipment: 23-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Ready-to-Use Dialysis Sacks
MWCO 12 to 14 kDa
Membrane Spectra/Por 1
Width (Metric) 50 mm
Capacity (Metric) 50 to 400 mL